Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC38A10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554713
Description
Immunogen sequence: MKPKQVSRDL GLAADLPGGA EGAAAQPQAV LRQPELRVIS DGEQGGQQGH RLDHGGHLEM RKA Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 31%.
SLC38A10 is a putative sodium-dependent amino acid/proton antiporter.
Specifications
SLC38A10 | |
Polyclonal | |
Unconjugated | |
SLC38A10 | |
1810073N04Rik; novel amino acid transporter protein; PP1744; Putative sodium-coupled neutral amino acid transporter 10; RGD1306356; SLC38A10; Solute carrier family 38 member 10; solute carrier family 38, member 10 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
124565 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HBR0 | |
SLC38A10 | |
Recombinant protein corresponding to Human SLC38A10. Recombinant protein control fragment (Product #RP-93480). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction