Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC38A10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542654
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542654 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542654 Supplier Invitrogen™ Supplier No. PA542654
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: HKEIHQQRQE DEEDKPRQVE VHQEPGAAVP RGQEAPEGKA RETVENLPPL Sequence homology: Guinea Pig: 93%; Human: 100%; Mouse: 91%; Rat: 93%; Yeast: 77%.

SLC38A10 is a putative sodium-dependent amino acid/proton antiporter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC38A10
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC38A10
Gene Accession No. Q9HBR0
Gene Alias 1810073N04Rik; novel amino acid transporter protein; PP1744; Putative sodium-coupled neutral amino acid transporter 10; RGD1306356; SLC38A10; Solute carrier family 38 member 10; solute carrier family 38, member 10
Gene Symbols SLC38A10
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human SLC38A10 (aa 730-779).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124565
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.