Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC38A2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113241
Description
SLC38A2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Under hypertonic conditions the induction of SLC38A2/SNAT2 leads to the stimulation of transport system A and to the increase in the cell content of amino acids. Its amino acid response element, along with a nearby conserved CAAT box, has enhancer activity in that it functions in an orientation and position independent manner, and it confers regulated transcription to a heterologous promoter.
Specifications
SLC38A2 | |
Polyclonal | |
Unconjugated | |
SLC38A2 | |
5033402L14Rik; AI316867; amino acid system A transporter; amino acid transporter 2; amino acid transporter A2; amino acid transporter cationic 2; amino acid transporter system A2; amino acid transporter, cationic 2 (low affinity); ATA2; Atrc2; Kiaa1382; mKIAA1382; PRO1068; Protein 40-9-1; Sa1; Sat2; SLC38A2; Snat2; sodium-coupled neutral amino acid transporter 2; Solute carrier family 38 member 2; solute carrier family 38, member 2; system A amino acid transporter 2; System A transporter 1; system N amino acid transporter 2 | |
Rabbit | |
Protein G | |
RUO | |
54407 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q96QD8 | |
SLC38A2 | |
Recombinant Human Sodium-coupled neutral amino acid transporter 2 protein (1-76AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction