Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC38A2/SNAT2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981958
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1958 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1958 Supplier Bioss Supplier No. BS12125R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Under hypertonic conditions the induction of SLC38A2/SNAT2 leads to the stimulation of transport system A and to the increase in the cell content of amino acids. Its amino acid response element, along with a nearby conserved CAAT box, has enhancer activity in that it functions in an orientation and position independent manner, and it confers regulated transcription to a heterologous promoter.

Specifications

Antigen SLC38A2/SNAT2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SLC38A2
Gene Accession No. Q96QD8
Gene Alias 5033402L14Rik; AI316867; amino acid system A transporter; amino acid transporter 2; Amino acid transporter A2; Ata2; KIAA1382; mKIAA1382; PRO1068; Protein 40-9-1; SAT2; SLC38A2; Snat2; Sodium-coupled neutral amino acid transporter 2; Solute carrier family 38 member 2; solute carrier family 38, member 2; System A amino acid transporter 2; System A transporter 1; System N amino acid transporter 2
Gene Symbols SLC38A2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SLC38A2/SNAT2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54407
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.