Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC38A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542143
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542143 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542143 Supplier Invitrogen™ Supplier No. PA542143
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: GNQRVEDPAR SCMEGKSFLQ KSPSKEPHFT DFEGKTSFGM SVFNLSNAIM Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Sheep: 86%.

As a sodium-dependent amino acid/proton antiporter, SLC38A3 mediates electrogenic cotransport of glutamine and sodium ions in exchange for protons. It also recognizes histidine, asparagine and alanine. It may mediate amino acid transport in either direction under physiological conditions and play a role in nitrogen metabolism and synaptic transmission.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC38A3
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC38A3
Gene Accession No. Q99624, Q9DCP2
Gene Alias 0610012J02Rik; D9Ucla2; G17; mNAT; N system amino acids transporter NAT-1; Na(+)-coupled neutral amino acid transporter 3; Na+ and H+ coupled amino acid transport system N1; NAT1; N-system amino acid transporter 1; RGD:628620}; Slc38-3; Slc38a3; slc38a3 {ECO:0000312; SLC38A3 protein; Sn1; Snat3; sodium-coupled neutral amino acid transporter 3; Solute carrier family 38 member 3; solute carrier family 38, member 3; system N amino acid transporter 1; system N1 amino acid transporter; system N1 Na+ and H+-coupled glutamine transporter
Gene Symbols SLC38A3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SLC38A3 (aa 31-80).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10991, 76257
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.