Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC38A3 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA549591

Catalog No. PA549591


Only null left
Add to Cart

Description

Description

SLC38A3 Polyclonal Antibody for Western Blot, Flow

As a sodium-dependent amino acid/proton antiporter, SLC38A3 mediates electrogenic cotransport of glutamine and sodium ions in exchange for protons. It also recognizes histidine, asparagine and alanine. It may mediate amino acid transport in either direction under physiological conditions and play a role in nitrogen metabolism and synaptic transmission.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SLC38A3
Polyclonal
Unconjugated
SLC38A3
0610012J02Rik; D9Ucla2; G17; mNAT; N system amino acids transporter NAT-1; Na(+)-coupled neutral amino acid transporter 3; Na+ and H+ coupled amino acid transport system N1; NAT1; N-system amino acid transporter 1; RGD:628620}; Slc38-3; Slc38a3; slc38a3 {ECO:0000312; SLC38A3 protein; Sn1; Snat3; sodium-coupled neutral amino acid transporter 3; Solute carrier family 38 member 3; solute carrier family 38, member 3; system N amino acid transporter 1; system N1 amino acid transporter; system N1 Na+ and H+-coupled glutamine transporter
Rabbit
Ammonium sulfate precipitation
RUO
10991
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Western Blot
2 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q99624
SLC38A3
KLH conjugated synthetic peptide between 442-468 amino acids from the Central region of human SLC38A3.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.