Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC38A3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592228
Description
Immunogen sequence: MEAPLQTEMV ELVPNGKHSE GLLPVITPMA GNQRVEDPAR SCMEGKSFLQ KSPSKEPHFT DFEGK; Positive Samples: HeLa, Raji, Mouse brain; Cellular Location: Cell membrane, Multi-pass membrane protein.
As a sodium-dependent amino acid/proton antiporter, SLC38A3 mediates electrogenic cotransport of glutamine and sodium ions in exchange for protons. It also recognizes histidine, asparagine and alanine. It may mediate amino acid transport in either direction under physiological conditions and play a role in nitrogen metabolism and synaptic transmission.
Specifications
SLC38A3 | |
Polyclonal | |
Unconjugated | |
SLC38A3 | |
0610012J02Rik; D9Ucla2; G17; mNAT; N system amino acids transporter NAT-1; Na(+)-coupled neutral amino acid transporter 3; Na+ and H+ coupled amino acid transport system N1; NAT1; N-system amino acid transporter 1; RGD:628620}; Slc38-3; Slc38a3; slc38a3 {ECO:0000312; SLC38A3 protein; Sn1; Snat3; sodium-coupled neutral amino acid transporter 3; Solute carrier family 38 member 3; solute carrier family 38, member 3; system N amino acid transporter 1; system N1 amino acid transporter; system N1 Na+ and H+-coupled glutamine transporter | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10991, 252919, 76257 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q99624, Q9DCP2, Q9JHZ9 | |
SLC38A3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-65 of human SLC38A3 (NP_0068321). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction