Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC38A4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542192
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542192 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542192 Supplier Invitrogen™ Supplier No. PA542192
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LAALFGYLTF YGEVEDELLH AYSKVYTLDI PLLMVRLAVL VAVTLTVPIV Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 92%; Horse: 92%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 92%; Rat: 100%; Zebrafish: 90%.

SLC38A4 is found predominantly in liver and transports both cationic and neutral amino acids. The transport of cationic amino acids by SLC38A4 is Na(+) and pH independent, while the transport of neutral amino acids is Na(+) and pH dependent.The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. Four alternatively spliced transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC38A4
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC38A4
Gene Accession No. Q969I6
Gene Alias 1110012E16Rik; 1700012A18Rik; amino acid transport system A3; amino acid transporter A3; amino acid transporter system A3; Ata3; FLJ10191; mATA3; MGC126876; mNAT3; N amino acid transporter 3; Na(+)-coupled neutral amino acid transporter 4; NAT3; PAAT; Slc38a4; Snat4; sodium-coupled neutral amino acid transporter 4; Solute carrier family 38 member 4; solute carrier family 38, member 4; system A amino acid transporter 3; System N amino acid transporter 3; system-N amino acid transporter 3; Unknown (protein for MGC:113830); wu:fd51c05; zgc:103694; zgc:113830
Gene Symbols SLC38A4
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human SLC38A4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55089
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.