Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC38A5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542504
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542504 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542504 Supplier Invitrogen™ Supplier No. PA542504
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: GIRAYEQLGQ RAFGPAGKVV VATVICLHNV GAMSSYLFII KSELPLVIGT Sequence homology: Cow: 93%; Dog: 86%; Guinea Pig: 85%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 100%; Rat: 100%.

The protein encoded by this gene is a system N sodium-coupled amino acid transporter involved in the transfer of glutamine, asparagine, histidine, serine, alanine, and glycine. The encoded protein does not transport charged amino acids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC38A5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC38A5
Gene Accession No. Q8WUX1
Gene Alias amino acid transport system N2; amino acid transporter system N2; C81234; E330031E14; JM24; PP7194; SLC38A5; Sn2; Snat5; sodium-coupled neutral amino acid transporter 5; sodium-coupled neutral amino acid transporter 5; LOW QUALITY PROTEIN: sodium-coupled neutral amino acid transporter 5; solute carrier family 38 (amino acid transporter), member 5; solute carrier family 38 member 5; solute carrier family 38, member 5; system N transporter 2; transport system N, protein 2
Gene Symbols SLC38A5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SLC38A5 (aa 107-156).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92745
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.