Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC38A6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$254.42 - $666.47
Specifications
| Antigen | SLC38A6 |
|---|---|
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
SLC38A6 Polyclonal specifically detects SLC38A6 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| SLC38A6 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| N system amino acid transporter NAT-1, SNAT6, solute carrier family 38, member 6 | |
| SLC38A6 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 145389 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:KWSIPCPLTLNYVEKGFQISNVTDDCKPKLFHFSKESA | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title