Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC38A6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113123
Description
SLC38A6 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
Specifications
| SLC38A6 | |
| Polyclonal | |
| Unconjugated | |
| SLC38A6 | |
| AW322671; EG625098; N system amino acid transporter NAT-1; Na(+)-coupled neutral amino acid transporter 6; NAT1; NAT-1; N-system amino acid transporter 1; Probable sodium-coupled neutral amino acid transporter 6; Slc38a6; Snat6; Solute carrier family 38 member 6; solute carrier family 38, member 6 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 145389, 625098 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| G3UVW3, Q8IZM9 | |
| SLC38A6 | |
| Recombinant Human Probable sodium-coupled neutral amino acid transporter 6 protein (132-169AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction