Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC39A11 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP25822425UL
Description
SLC39A11 Polyclonal specifically detects SLC39A11 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
SLC39A11 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 1-4 μg/mL | |
C17orf26, chromosome 17 open reading frame 26, solute carrier family 39 (metal ion transporter), member 11, Solute carrier family 39 member 11, zinc transporter ZIP11, ZIP11, ZIP-11, Zrt- and Irt-like protein 11 | |
Rabbit | |
Affinity Purified | |
RUO | |
201266 | |
Human | |
IgG |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide | |
SLC39A11 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:TASATFESARNLAIGIGIQNFPEGLAVSLPLRGAGFSTWRAFW | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction