Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC39A2/ZIP2 Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176499MFV450
Description
SLC39A2/ZIP2 Polyclonal antibody specifically detects SLC39A2/ZIP2 in Human, Mouse, Rat, Equine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
SLC39A2/ZIP2 | |
Polyclonal | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
Eti-1, MGC119190, solute carrier family 39 (zinc transporter), member 2, Solute carrier family 39 member 2, zinc transporter ZIP2, ZIP-2, ZIP2hZIP2, Zrt- and Irt-like protein 2,6A1 | |
Antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human ZIP2. The immunogen is located within amino acids 70 - 120 of ZIP2. Amino Acid Squence: NRSASERNSSGDADSAH | |
0.1 mL | |
Cancer, Cell Cycle and Replication | |
29986 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Equine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction