Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC39A3 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176500PCP
Description
SLC39A3 Polyclonal antibody specifically detects SLC39A3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
SLC39A3 | |
Polyclonal | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
solute carrier family 39 (zinc transporter), member 3, zinc transporter ZIP3, ZIP-3, ZIP3Solute carrier family 39 member 3, Zrt- and Irt-like protein 3 | |
Antibody was raised against a 16 amino acid synthetic peptide near the center of human ZIP3. The immunogen is located within amino acids 110 - 160 of ZIP3. Amino Acid Squence: NAGSDVGSDSEYESPF | |
0.1 mL | |
Cancer, Cell Cycle and Replication | |
29985 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction