Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC39A4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547915
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547915 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547915 Supplier Invitrogen™ Supplier No. PA547915
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.

The zinc transporter ZIP4, also known as SLC39A4, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. ZIP4 was initially identified as zinc-uptake transporter in the intestine and as a candidate gene for acrodermatitis enteropathica (AE), a rare inherited defect in the absorption of dietary zinc. Similar to other ZIP family members, aberrant expression of ZIP4 has been linked to cancer, specifically the progression and aggressiveness of pancreatic cancer. ZIP4 expression has also been shown to be activated in hepatocellular carcinomas, acting to repress apoptosis, enhance cell cycle and increase migration, suggesting ZIP4 may play a role in multiple types of cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC39A4
Applications Flow Cytometry, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Slc39a4
Gene Accession No. Q78IQ7
Gene Alias 1600025H15Rik; activated in W/Wv mouse stomach 2; AEZ; AU041686; AWMS2; EMBL:AAI27515.1}; mAWMS2; SLC39A4; slc39a4 {ECO:0000312; solute carrier family 39 (zinc transporter), member 4; solute carrier family 39 member 4; Zinc transporter ZIP4; Zip4; ZIP-4; Zrt- and Irt-like protein 4
Gene Symbols Slc39a4
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse SLC39A4 Met1-Tyr337.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 72027
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.