Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC39A5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542522
Description
Peptide sequence: MGSPVSHLLA GFCVWVVLGW VGGSVPNLGP AEQEQNHYLA QLFGLYGENG Sequence homology: Cow: 86%; Dog: 93%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
The zinc transporter ZIP5, also known as SLC39A5, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. ZIP5 is closely related to ZIP4, and is found in many of the same tissues, including intestine, pancreas, liver, and kidney, but unlike ZIP4, ZIP5 localizes to the basolateral membrane in kidney cells and its activity is not downregulated by zinc repletion. Recent evidence suggests that ZIP4 and ZIP5 are reciprocally regulated, with translation of ZIP5 mRNA decreasing and ZIP5 protein being rapidly internalized and degraded in response to zinc deficiency.
Specifications
SLC39A5 | |
Polyclonal | |
Unconjugated | |
SLC39A5 | |
1810013D05Rik; 2010205A06Rik; LZT-Hs7; MGC34778; MYP24; SLC39A5; solute carrier family 39 (metal ion transporter), member 5; solute carrier family 39 (zinc transporter), member 5; solute carrier family 39 member 5; Zinc transporter ZIP5; Zip5; ZIP-5; zrt- and Irt-like protein 5 | |
Rabbit | |
Affinity chromatography | |
RUO | |
283375 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6ZMH5 | |
SLC39A5 | |
Synthetic peptide directed towards the N-terminal of human SLC39A5 (aa 2-51). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction