Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC39A5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542523
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542523 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542523 Supplier Invitrogen™ Supplier No. PA542523
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: HYLAQLFGLY GENGTLTAGG LARLLHSLGL GRVQGLRLGQ HGPLTGRAAS Sequence homology: Cow: 86%; Dog: 86%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%.

The zinc transporter ZIP5, also known as SLC39A5, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. ZIP5 is closely related to ZIP4, and is found in many of the same tissues, including intestine, pancreas, liver, and kidney, but unlike ZIP4, ZIP5 localizes to the basolateral membrane in kidney cells and its activity is not downregulated by zinc repletion. Recent evidence suggests that ZIP4 and ZIP5 are reciprocally regulated, with translation of ZIP5 mRNA decreasing and ZIP5 protein being rapidly internalized and degraded in response to zinc deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC39A5
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC39A5
Gene Accession No. Q6ZMH5
Gene Alias 1810013D05Rik; 2010205A06Rik; LZT-Hs7; MGC34778; MYP24; SLC39A5; solute carrier family 39 (metal ion transporter), member 5; solute carrier family 39 (zinc transporter), member 5; solute carrier family 39 member 5; Zinc transporter ZIP5; Zip5; ZIP-5; zrt- and Irt-like protein 5
Gene Symbols SLC39A5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SLC39A5 (aa 38-87).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283375
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.