Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC39A6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533036
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533036 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533036 Supplier Thermo Scientific Supplier No. PA533036
Add to Cart
Add to Cart

SLC39A6 Polyclonal Antibody

SLC39A6 Polyclonal Antibody for IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC39A6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene SLC39A6
Gene Accession No. Q13433
Gene Alias EMBL:AAH97493.1}; endoplasmic reticulum membrane protein; endoplasmic reticulum membrane-linked protein; Ermelin; Estrogen-regulated protein LIV-1; LIV1; LIV-1; LIV-1 protein, estrogen regulated; SLC39A6; slc39a6 {ECO:0000312; solute carrier family 39 (metal ion transporter), member 6; solute carrier family 39 (zinc transporter), member 6; solute carrier family 39 member 6; Zinc transporter ZIP6; Zip6; ZIP-6; zrt- and Irt-like protein 6
Gene Symbols SLC39A6
Host Species Rabbit
Immunogen Synthetic 20 amino acid peptide from internal region of human SLC39A6.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25800
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.