Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC39A6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583605
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583605 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583605 Supplier Invitrogen™ Supplier No. PA583605
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HSHHNHAASG KNKRKALCPD HDSDSSGKDP RNSQGKGAHR PEHASGRRNV KDSVSASEVT STVYNTVSEG THFLETIETP RPGKLFPKDV SSSTPPSVTS KSRVSRLAGR KTNESVSEPR KGFMYSRNTN ENPQE Highest antigen sequence indentity to the following orthologs: Rat - 70%, Mouse - 68%.

This gene encodes a protein which interacts with the carboxy-terminus of transcription initiation factor TFIIF, a transcription factor which regulates elongation as well as initiation by RNA polymerase II. The protein may also represent a component of an RNA polymerase II holoenzyme complex. Alternative splicing of this gene results in two transcript variants encoding 2 different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC39A6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC39A6
Gene Accession No. Q13433
Gene Alias EMBL:AAH97493.1}; endoplasmic reticulum membrane protein; endoplasmic reticulum membrane-linked protein; Ermelin; Estrogen-regulated protein LIV-1; LIV1; LIV-1; LIV-1 protein, estrogen regulated; SLC39A6; slc39a6 {ECO:0000312; solute carrier family 39 (metal ion transporter), member 6; solute carrier family 39 (zinc transporter), member 6; solute carrier family 39 member 6; Zinc transporter ZIP6; Zip6; ZIP-6; zrt- and Irt-like protein 6
Gene Symbols SLC39A6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC39A6. Recombinant protein control fragment (Product #RP-91283).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25800
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.