Learn More
Invitrogen™ SLC39A6 Polyclonal Antibody

Description
A suggested positive control is mouse lung tissue lysate. PA5-21071 can be used with blocking peptide PEP-1185.
Specifications
Specifications
| Antigen | SLC39A6 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | SLC39A6 |
| Gene Accession No. | Q13433, Q8C145 |
| Gene Alias | EMBL:AAH97493.1}; endoplasmic reticulum membrane protein; endoplasmic reticulum membrane-linked protein; Ermelin; Estrogen-regulated protein LIV-1; LIV1; LIV-1; LIV-1 protein, estrogen regulated; SLC39A6; slc39a6 {ECO:0000312; solute carrier family 39 (metal ion transporter), member 6; solute carrier family 39 (zinc transporter), member 6; solute carrier family 39 member 6; Zinc transporter ZIP6; Zip6; ZIP-6; zrt- and Irt-like protein 6 |
| Gene Symbols | SLC39A6 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.