Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC39A7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562893
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562893 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562893 Supplier Invitrogen™ Supplier No. PA562893
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GHDDLHDDLQ EDFHGHSHRH SHEDFHHGHS HAHGHGHTHE SIWHGHTHDH DHGHSHEDLH HG Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 82%.

The zinc transporter ZIP7, also known as SLC39A7, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. ZIP7 was initially identified while characterizing genes in the major histocompatibility complex on chromosome 17. ZIP7 mRNA is abundantly and widely expressed and the protein localizes to the Golgi apparatus. It functions to transport intracellular zinc from the Golgi apparatus to the cytoplasm of the cell. ZIP7 expression is expressed by zinc. ZIP7 has been suggested to act a hub for tyrosine kinase activation and may thus be a potential therapeutic target for diseases such as cancer where prevention of tyrosine kinase activation would be advantageous.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC39A7
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC39A7
Gene Accession No. Q92504
Gene Alias AA408174; AI117660; AL024048; D6S115E; D6S2244E; DADB-100D22.3; H2-K region expressed gene 4; H-2Ke4; H2-KE4; Histidine-rich membrane protein; histidine-rich membrane protein Ke4; HKE4; HLA class II region expressed gene KE4; KE4; Ke-4; Ke4 gene; Ke4 gene, mouse, human homolog of; really interesting new gene 5 protein; Ring5; RT1 class I, locus Ke4; RT1 region expressed gene KE4; Rt1Ke4; RT1-Ke4; Slc39a7; solute carrier family 39 (zinc transporter), member 7; solute carrier family 39 member 7; Unknown (protein for MGC:142379); zinc transporter SLC39A7; ZIP7; zrt-, Irt-like protein 7
Gene Symbols SLC39A7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC39A7. Recombinant protein control fragment (Product #RP-101736).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7922
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.