Learn More
SLC39A8 Polyclonal Antibody

Description
SLC39A8 Polyclonal Antibody for Western Blot, Flow
Specifications
Specifications
| Antigen | SLC39A8 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | Slc39a8 |
| Gene Accession No. | Q9C0K1 |
| Gene Alias | 4933419D20Rik; AA986696; BCG induced integral membrane protein BIGM103; BCG-induced integral membrane protein in monocyte clone 103 protein; BIGM103; CDG2N; LIV-1 subfamily of ZIP zinc transporter 6; LZT-Hs6; PP3105; Slc39a8; solute carrier family 39 (metal ion transporter), member 8; solute carrier family 39 (zinc transporter), member 8; solute carrier family 39 (zinc transporter), member 8 tv2; solute carrier family 39 member 8; Zinc transporter ZIP8; Zip8; ZIP-8; Zrt- and Irt-like protein 8 |
| Gene Symbols | Slc39a8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.