Learn More
Invitrogen™ SLC39A8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587865
Description
Immunogen sequence: QLIPEAFGFD PKVDSYVEKA VAVFGGFYLL FFFERMLKML LKTYGQNGHT HFGNDNFGPQ EKTHQPKALP AINGVTCYAN PAVTEANGHI HFDNVSVVSL QDGKKEPSSC TCLKGPKLSE I; Positive Samples: HL-60, NCI-H460, HeLa, Mouse liver; Cellular Location: Membrane, Multi-pass membrane protein.
The zinc transporter ZIP8, also known as SLC39A9, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. The zinc transporter family is divided into four subfamilies (I, II, LIV-1 and gufA). ZIP8 is glycosylated and located at the plasma membrane and mitochondria. It has been identified as the transporter responsible for transport of the toxic Cadmium cation. ZIP8 has also been suggested to play a role in the regulation of interferon-gamma expression in activated human T cells.
Specifications
SLC39A8 | |
Polyclonal | |
Unconjugated | |
Slc39a8 | |
4933419D20Rik; AA986696; BCG induced integral membrane protein BIGM103; BCG-induced integral membrane protein in monocyte clone 103 protein; BIGM103; CDG2N; LIV-1 subfamily of ZIP zinc transporter 6; LZT-Hs6; PP3105; Slc39a8; solute carrier family 39 (metal ion transporter), member 8; solute carrier family 39 (zinc transporter), member 8; solute carrier family 39 (zinc transporter), member 8 tv2; solute carrier family 39 member 8; Zinc transporter ZIP8; Zip8; ZIP-8; Zrt- and Irt-like protein 8 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
64116, 67547 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q91W10, Q9C0K1 | |
Slc39a8 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 180-300 of human ZIP8 (NP_0714373). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.