Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC39A8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102899
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102899 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102899 Supplier Invitrogen™ Supplier No. PA5102899
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total BIGM103.

The zinc transporter ZIP8, also known as SLC39A9, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. The zinc transporter family is divided into four subfamilies (I, II, LIV-1 and gufA). ZIP8 is glycosylated and located at the plasma membrane and mitochondria. It has been identified as the transporter responsible for transport of the toxic Cadmium cation. ZIP8 has also been suggested to play a role in the regulation of interferon-gamma expression in activated human T cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC39A8
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Slc39a8
Gene Accession No. Q9C0K1
Gene Alias 4933419D20Rik; AA986696; BCG induced integral membrane protein BIGM103; BCG-induced integral membrane protein in monocyte clone 103 protein; BIGM103; CDG2N; LIV-1 subfamily of ZIP zinc transporter 6; LZT-Hs6; PP3105; Slc39a8; solute carrier family 39 (metal ion transporter), member 8; solute carrier family 39 (zinc transporter), member 8; solute carrier family 39 (zinc transporter), member 8 tv2; solute carrier family 39 member 8; Zinc transporter ZIP8; Zip8; ZIP-8; Zrt- and Irt-like protein 8
Gene Symbols Slc39a8
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLC39A8(Accession Q9C0K1), corresponding to amino acid residues S94-L144.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64116
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.