Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC39A9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101970
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101970 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101970 Supplier Invitrogen™ Supplier No. PA5101970
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SLC39A9.

The zinc transporter ZIP9, also known as SLC39A9, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. The zinc transporter family is divided into four subfamilies (I, II, LIV-1 and gufA). ZIP9 is a multipass membrane protein that belongs to the ZIP transporter subfamily I. It is located at the trans-Golgi network regardless of zinc presence and is thought to be a zinc homeostasis regulator acting in the secretory pathway without significantly altering cytosolic zinc homeostasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC39A9
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SLC39A9
Gene Accession No. Q3KR82, Q9NUM3
Gene Alias 2010002A02Rik; 2610511I23Rik; 4833420E20Rik; AW209151; SLC39A9; solute carrier family 39 (zinc transporter), member 9; solute carrier family 39 member 9; solute carrier family 39, member 9; UNQ714/PRO1377; zinc transporter SLC39A9; zinc transporter ZIP9; ZIP9; ZIP-9; Zrt- and Irt-like protein 9
Gene Symbols SLC39A9
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLC39A9(Accession Q9NUM3), corresponding to amino acid residues G109-D159.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 314275, 55334
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.