Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC3A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542409
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542409 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542409 Supplier Invitrogen™ Supplier No. PA542409
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: DFREVDPIFG TMEDFENLVA AIHDKGLKLI IDFIPNHTSD KHIWFQLSRT Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.

This gene encodes a type II membrane glycoprotein which is one of the components of the renal amino acid transporter which transports neutral and basic amino acids in the renal tubule and intestinal tract.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC3A1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC3A1
Gene Accession No. Q07837
Gene Alias amino acid transport related protein; amino acid transporter 1; amino acid transporter SLC3A1; ATR1; b(0,+)-type amino acid transport protein; CSNU1; D2; D2H; FLJ34681; membrane glycoprotein; NAA-TR; Nbat; Neutral and basic amino acid transport protein rBAT; NTAA; rBAT; rBAT protein; si:dkey-202m23.2; Slc3a1; SLC3A1 variant B; SLC3A1 variant C; SLC3A1 variant D; SLC3A1 variant E; SLC3A1 variant F; SLC3A1 variant G; solute carrier family 3 (amino acid transporter heavy chain), member 1; solute carrier family 3 (cystine, dibasic and neutral amino acid transporters), member 1; solute carrier family 3 (cystine, dibasic and neutral amino acid transporters, activator of cystine, dibasic and neutral amino acid transport), member 1; solute carrier family 3 member 1; solute carrier family 3, member 1
Gene Symbols SLC3A1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SLC3A1 (aa 179-228).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6519
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.