Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC41A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117339
Description
Centrifuge briefly prior to opening.
SLC41A1 belongs to the SLC41A transporter family. It acts as a magnesium transporter that is responsive to magnesium balance.
Specifications
SLC41A1 | |
Polyclonal | |
Unconjugated | |
SLC41A1 | |
AI573938; B230315F01Rik; MgtE; Slc41a1; solute carrier family 41 (magnesium transporter), member 1; solute carrier family 41 member 1; solute carrier family 41, member 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
254428, 98396 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.74 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q8BJA2, Q8IVJ1 | |
SLC41A1 | |
Recombinant protein encompassing a sequence within the C-terminus region of human SLC41A1. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction