Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC41A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107266
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107266 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107266 Supplier Invitrogen™ Supplier No. PA5107266
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence homology: Cow: 92%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 92%.

SLC41A1 belongs to the SLC41A transporter family. It acts as a magnesium transporter that is responsive to magnesium balance.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC41A1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene SLC41A1
Gene Accession No. Q8IVJ1
Gene Alias AI573938; B230315F01Rik; MgtE; Slc41a1; solute carrier family 41 (magnesium transporter), member 1; solute carrier family 41 member 1; solute carrier family 41, member 1
Gene Symbols SLC41A1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human SLC41A1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 254428
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.