Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC43A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537059
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537059 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537059 Supplier Invitrogen™ Supplier No. PA537059
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 61 kDa. Purity is >95% by SDS-PAGE.

SLC43A1 belongs to the system L family of plasma membrane carrier proteins that transports large neutral amino acids. SLC43A1 belongs to the system L family of plasma membrane carrier proteins that transports large neutral amino acids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC43A1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC43A1
Gene Accession No. O75387, Q8BSM7
Gene Alias 2610016F07Rik; AA986141; large neutral amino acids transporter small subunit 3; LAT3; L-type amino acid transporter 3; PB39; POV1; prostate cancer overexpressed gene 1; prostate cancer overexpressed gene 1 protein; R00504; Slc43a1; solute carrier family 43 (amino acid system L transporter), member 1; solute carrier family 43 member 1; solute carrier family 43, member 1
Gene Symbols SLC43A1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 275-320 of human SLC43A1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 311168, 72401, 8501
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.