Learn More
SLC43A1 Rabbit anti-Human, Polyclonal, ProteinTech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 104441AP150UL

Description
Aliquoting is unnecessary for -20°C storage.
SLC43A1 Polyclonal antibody specifically detects SLC43A1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
SLC43A1 | |
Polyclonal | |
Unconjugated | |
SLC43A1 | |
2610016F07Rik; AA986141; large neutral amino acids transporter small subunit 3; LAT3; L-type amino acid transporter 3; PB39; POV1; prostate cancer overexpressed gene 1; prostate cancer overexpressed gene 1 protein; R00504; Slc43a1; solute carrier family 43 (amino acid system L transporter), member 1; solute carrier family 43 member 1; solute carrier family 43, member 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8501 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
343 μg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
O75387 | |
SLC43A1 | |
SLC43A1 Fusion Protein Ag0305 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.