Learn More
Invitrogen™ SLC43A2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113783
Description
Immunogen sequence: TEPENVTNGT VGGTAEPGHE EVSWMNGWLS CQAQDEMLNL AFTVGSFLLS For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 79%; Dog: 93%; Horse: 85%; Human: 100%; Pig: 86%; Rat: 86%.
Uniporter that mediates the transport of the stereospecific L-phenylalanine, L-methionine and L-branched-chain amino acids, between the extracellular space and the cytoplasm and may control the transepithelial (re)absorption of neutral amino acid in kidney and small intestine. The transport activity is mediated through facilitated diffusion and is sodium ions-, chloride ions- and pH-independent.
Specifications
SLC43A2 | |
Polyclonal | |
Unconjugated | |
SLC43A2 | |
7630402D21Rik; BC042513; large neutral amino acids transporter small subunit 4; Lat4; L-type amino acid transporter 4; L-type amino L-acid transporter 4; PP7664; Slc43a2; solute carrier family 43 (amino acid system L transporter), member 2; solute carrier family 43 member 2; solute carrier family 43, member 2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
124935 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8N370 | |
SLC43A2 | |
Synthetic peptide directed towards the N terminal region of human SLC43A2 (aa 51-100). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.