Learn More
Invitrogen™ SLC43A2 Polyclonal Antibody

Description
This antibody is predicted to react with bovine based on sequence homology.
Specifications
Specifications
| Antigen | SLC43A2 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | SLC43A2 |
| Gene Accession No. | Q8CGA3, Q8N370 |
| Gene Alias | 7630402D21Rik; BC042513; large neutral amino acids transporter small subunit 4; Lat4; L-type amino acid transporter 4; L-type amino L-acid transporter 4; PP7664; Slc43a2; solute carrier family 43 (amino acid system L transporter), member 2; solute carrier family 43 member 2; solute carrier family 43, member 2 |
| Gene Symbols | SLC43A2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.