Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC43A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585024
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585024 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585024 Supplier Invitrogen™ Supplier No. PA585024
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FKNEDYFKDL CGPDAGPIGN ATGQADCKAQ DERFS Highest antigen sequence indentity to the following orthologs: Mouse - 51%, Rat - 54%.

SLC43A3 belongs to the SLC43A transporter family and is a putative transporter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC43A3
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC43A3
Gene Accession No. Q8NBI5
Gene Alias DKFZp762A227; EEG1; FOAP-13; likely ortholog of mouse embryonic epithelial gene 1; PRO1659; Protein FOAP-13; PSEC0252; SEEEG-1; SLC43A3; Solute carrier family 43 member 3; solute carrier family 43, member 3
Gene Symbols SLC43A3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC43A3. Recombinant protein control fragment (Product #RP-108676).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29015
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.