Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC43A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542460
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542460 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542460 Supplier Invitrogen™ Supplier No. PA542460
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MAGQGLPLHV ATLLTGLLEC LGFAGVLFGW PSLVFVFKNE DYFKDLCGPD Sequence homology: Dog: 92%; Horse: 86%; Human: 100%; Rabbit: 92%.

SLC43A3 belongs to the SLC43A transporter family and is a putative transporter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC43A3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC43A3
Gene Accession No. Q8NBI5
Gene Alias DKFZp762A227; EEG1; FOAP-13; likely ortholog of mouse embryonic epithelial gene 1; PRO1659; Protein FOAP-13; PSEC0252; SEEEG-1; SLC43A3; Solute carrier family 43 member 3; solute carrier family 43, member 3
Gene Symbols SLC43A3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SLC43A3 (aa 1-50).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29015
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.