Learn More
Invitrogen™ SLC44A1 Monoclonal Antibody (VIM-15b), PE
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA528191
Description
Mouse anti human CD92 antibody, clone VIM-15b recognizes the C-terminal variant of human CDw92 (isoform 3), also known as CTL1, a 70 kDa multi-pass membrane protein, expressed by monocytes, neutrophils, certain myeloid and T cell lines, and weakly by endothelial cells, fibroblasts and epithelial cells. Reconstitute by adding 1 mL distilled water.
SLC44A1 encodes a choline transporter and may be involved in membrane synthesis and myelin production.
Specifications
SLC44A1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 5% sucrose and 0.09% sodium azide | |
Q8WWI5 | |
Slc44a1 | |
MV4-11 acute monocyte leukemia cells. | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry | |
VIM-15b | |
PE | |
Slc44a1 | |
2210409B22Rik; 4833416H08Rik; AW547365; CD92; CDW92; CDW92 antigen; choline transporter-like protein 1; CHTL1; Ctl1; Slc44a1; solute carrier family 44 (choline transporter), member 1; solute carrier family 44 member 1; solute carrier family 44, member 1; transporter-like protein | |
Mouse | |
Protein G | |
RUO | |
23446 | |
4°C, store in dark, DO NOT FREEZE! | |
Lyophilized |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.