Learn More
Invitrogen™ SLC44A1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590449
Description
Immunogen sequence: AARLVSGYDS YGNICGQKNT KLEAIPNSGM DHTQRKYVFF LDPCNLDLIN RKIKSVALCV AACPRQELKT LSDVQKFAEI NGSALCSYNL KPSEYTTSPK SSVLCPKLPV PASAPIPFFH RCAPVNISCY AKFAEALITF VSDNSVLHRL ISGVMT; Positive Samples: HeLa, A-549, LO2, mouse liver, mouse brain, mouse kidney, rat liver; Cellular Location: Cell membrane, Mitochondrion outer membrane, Multi-pass membrane protein.
SLC44A1 encodes a choline transporter and may be involved in membrane synthesis and myelin production.
Specifications
SLC44A1 | |
Polyclonal | |
Unconjugated | |
Slc44a1 | |
2210409B22Rik; 4833416H08Rik; AW547365; CD92; CDW92; CDW92 antigen; choline transporter-like protein 1; CHTL1; Ctl1; Slc44a1; solute carrier family 44 (choline transporter), member 1; solute carrier family 44 member 1; solute carrier family 44, member 1; transporter-like protein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
100434, 23446, 85254 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q6X893, Q8VII6, Q8WWI5 | |
Slc44a1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 55-210 of human SLC44A1 (NP_5368562). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.