Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC44A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139986
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139986 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139986 Supplier Invitrogen™ Supplier No. PA5139986
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: LFLCLAIDTK YNDGSPGREF YMDKVLMEFV ENSRKAMKEA GKGGVADSRE LKPMASGASS Highest antigen sequence indentity to the following orthologs: Rat - 95%, Mouse - 95%.

SLC44A1 encodes a choline transporter and may be involved in membrane synthesis and myelin production.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC44A1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Slc44a1
Gene Accession No. Q8WWI5
Gene Alias 2210409B22Rik; 4833416H08Rik; AW547365; CD92; CDW92; CDW92 antigen; choline transporter-like protein 1; CHTL1; Ctl1; Slc44a1; solute carrier family 44 (choline transporter), member 1; solute carrier family 44 member 1; solute carrier family 44, member 1; transporter-like protein
Gene Symbols Slc44a1
Host Species Rabbit
Immunogen Recombinant protein fragment of Human SLC44A1 (Product #RP-107364)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23446
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.