Learn More
Invitrogen™ SLC44A2 Polyclonal Antibody

Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Orangutan, Gibbon, Monkey, Marmoset (100%); Galago (94%); Bat (88%); Rat, Hamster, Elephant, Panda, Dog (81%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
Specifications
Specifications
| Antigen | SLC44A2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | SLC44A2 |
| Gene Accession No. | Q8IWA5 |
| Gene Alias | 1110028E10Rik; Choline transporter-like protein 2; Ctl2; IESCA; inner ear supporting cell antigen; PP1292; PSEC0210; RGD1309680; SLC44A2; solute carrier family 44 (choline transporter), member 2; solute carrier family 44 member 2; solute carrier family 44, member 2; solute carrier family 44, member 2 tv2; testicular tissue protein Li 47 |
| Gene Symbols | SLC44A2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.