Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC44A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534205
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534205 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534205 Supplier Invitrogen™ Supplier No. PA534205
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla (100%); Gibbon, Monkey, Marmoset (94%); Orangutan, Galago, Dog (88%); Rat, Hamster, Elephant, Panda, Horse, Pig, Guinea pig (81%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

Isoform 1, but not isoform 3, exhibits some choline transporter activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC44A2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene SLC44A2
Gene Accession No. Q8IWA5
Gene Alias 1110028E10Rik; Choline transporter-like protein 2; Ctl2; IESCA; inner ear supporting cell antigen; PP1292; PSEC0210; RGD1309680; SLC44A2; solute carrier family 44 (choline transporter), member 2; solute carrier family 44 member 2; solute carrier family 44, member 2; solute carrier family 44, member 2 tv2; testicular tissue protein Li 47
Gene Symbols SLC44A2
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from internal region of human SLC44A2.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57153
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.