Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC44A3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561517
Description
Immunogen sequence: GAAGRLLFGY DSFGNMCGKK NSPVEGAPLS GQDMTLKKHV FFMNSCNLEV KGTQLNRMAL CVSNCPEEQL DSLEEVQ Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 84%.
SLC44A3 is a multi-pass membrane protein. It belongs to the CTL (choline transporter-like) family. The function of the RBM34 protein remains unknown.
Specifications
SLC44A3 | |
Polyclonal | |
Unconjugated | |
SLC44A3 | |
BC010552; choline transporter-like protein 3; Ctl3; RGD1305808; SLC44A3; Solute carrier family 44 member 3; solute carrier family 44, member 3; UNQ558/PRO1115 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
126969 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N4M1 | |
SLC44A3 | |
Recombinant protein corresponding to Human SLC44A3. Recombinant protein control fragment (Product #RP-99788). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction