Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC44A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588779
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588779 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588779 Supplier Invitrogen™ Supplier No. PA588779
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLLFGYDSFG NMCGKKNSPV EGAPLSGQDM TLKKHVFFMN SCNLEVKGTQ LNRMALCVSN CPEEQLDSLE EVQFFANTSG SFLCVYSLNS FNYTHSPKAD SLCPRLPVPP SKSFPLFNRC VPQTPECYSL FASVLINDVD TLHRILSGIM SGRDTILGLC I; Positive Samples: HT-29, U-87MG, HeLa; Cellular Location: Membrane, Multi-pass membrane protein.

SLC44A3 is a multi-pass membrane protein. It belongs to the CTL (choline transporter-like) family. The function of the RBM34 protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC44A3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SLC44A3
Gene Accession No. Q8N4M1
Gene Alias BC010552; choline transporter-like protein 3; Ctl3; RGD1305808; SLC44A3; Solute carrier family 44 member 3; solute carrier family 44, member 3; UNQ558/PRO1115
Gene Symbols SLC44A3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 60-220 of human SLC44A3 (NP_0011075781).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 126969
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.