Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC44A4 Polyclonal Antibody
SLC44A4 Polyclonal Antibody
Supplier: Thermo Scientific PA561391
Description
SLC44A4 Polyclonal Antibody for IHC

Specifications
SLC44A4 | |
Polyclonal | |
Unconjugated | |
SLC44A4 | |
C6orf29; choline transporter-like protein 4; CTL4; hTPPT1; NG22; SLC44A4; Solute carrier family 44 member 4; solute carrier family 44, member 4; testicular tissue protein Li 48; thiamine pyrophosphate transporter; Thiamine pyrophosphate transporter 1; TPPT; TPPT1; UNQ441/PRO874 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
80736 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q53GD3 | |
SLC44A4 | |
Recombinant protein corresponding to Human SLC44A4. Recombinant protein control fragment (Product #RP-99593). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction