Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC45A2 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA542416
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542416 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542416 Supplier Thermo Scientific Supplier No. PA542416

Rabbit Polyclonal Antibody

Peptide sequence: IGWTAFLSNM LFFTDFMGQI VYRGDPYSAH NSTEFLIYER GVEVGCWGFC Sequence homology: Cow: 86%; Dog: 93%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 79%; Rat: 100%

SLC45A2 Polyclonal antibody specifically detects SLC45A2 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC45A2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC45A2
Gene Accession No. Q9UMX9
Gene Alias 1A1; AIM1; Aim-1; B/AIM-1-like protein; blanc-sale; bls; Dbr; dominant brown; Matp; Melanoma antigen AIM1; membrane associated transport protein; membrane associated transporter protein; membrane-associated transporter protein; Oca4; protein AIM-1; Protein underwhite; SHEP5; Slc45a2; Solute carrier family 45 member 2; solute carrier family 45, member 2; underwhite; uw
Gene Symbols SLC45A2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human SLC45A2
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51151
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.