Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC45A4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
| Antigen | SLC45A4 |
|---|---|
| Concentration | LYOPH |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
SLC45A4 Polyclonal specifically detects SLC45A4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| SLC45A4 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| 57210 | |
| IgG | |
| Unpurified | |
| Human. Other species not yet tested. |
| LYOPH | |
| Polyclonal | |
| Antisera | |
| RUO | |
| KIAA1126, solute carrier family 45, member 4 | |
| SLC45A4 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title