Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC46A1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569283
Description
This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Has been shown to act both as an intestinal proton-coupled high-affinity folate transporter and as an intestinal heme transporter which mediates heme uptake from the gut lumen into duodenal epithelial cells. The iron is then released from heme and may be transported into the bloodstream. Dietary heme iron is an important nutritional source of iron. Shows a higher affinity for folate than heme.
Specifications
SLC46A1 | |
Polyclonal | |
Unconjugated | |
SLC46A1 | |
1110002C08Rik; D11Ertd18e; G21; Hcp1; Heme carrier protein 1; Pcft; PCFT/HCP1; proton-coupled folate transporter; retinal pigment epithelilum transporter; retinal pigment epithelium transporter; RGD1309472; Slc46a1; solute carrier family 46 (folate transporter), member 1; solute carrier family 46 member 1; solute carrier family 46, member 1; TRPE; unnamed protein product | |
Rabbit | |
Affinity chromatography | |
RUO | |
113235 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96NT5 | |
SLC46A1 | |
synthetic peptide directed towards the N terminal region of human SLC46A1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction