Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC46A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598572
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598572 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598572 Supplier Invitrogen™ Supplier No. PA598572
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC46A3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

The function remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC46A3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SLC46A3
Gene Accession No. Q7Z3Q1
Gene Alias 1200006F02Rik; FKSG16; RGD1307594; SLC46A3; Solute carrier family 46 member 3; solute carrier family 46, member 3
Gene Symbols SLC46A3
Host Species Rabbit
Immunogen Recombinant Human Solute carrier family 46 member 3 protein (26-73AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283537
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.