Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC46A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542530
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542530 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542530 Supplier Invitrogen™ Supplier No. PA542530
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MKILFVEPAI FLSAFAMTLT GPLTTQYVYR RIWEETGNYT FSSDSNISEC Sequence homology: Dog: 79%; Human: 100%; Mouse: 85%; Rabbit: 79%; Rat: 79%.

The function remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC46A3
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC46A3
Gene Accession No. Q7Z3Q1
Gene Alias 1200006F02Rik; FKSG16; RGD1307594; SLC46A3; Solute carrier family 46 member 3; solute carrier family 46, member 3
Gene Symbols SLC46A3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SLC46A3 (aa 1-50).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283537
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.