Learn More
Invitrogen™ SLC47A1 Polyclonal Antibody

Description
SLC47A1 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | SLC47A1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | Slc47a1 |
| Gene Accession No. | Q96FL8 |
| Gene Alias | 1300013J15Rik; AI036982; FLJ10847; H+/organic cation antiporter variant 1; H+/organic cation antiporter variant 2; hMATE-1; MATE1; MATE-1; MGC64822; mMATE1; mMATE-1; multidrug and toxin extrusion 1; multidrug and toxin extrusion protein 1; RGD1311123; rMATE-1; SLC47A1; solute carrier family 47 (multidrug and toxin extrusion), member 1; solute carrier family 47 member 1; solute carrier family 47, member 1 |
| Gene Symbols | Slc47a1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.