Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC47A1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589905
Description
Immunogen sequence: LNWKKACQQA QVHANLKVNN VPRSGNSALP QDPLHPGCPE NLEGILTNDV GKTGEPQSDQ QMRQEEPLPE HPQDGAKLSR KQLVLRRGLL L; Positive Samples: Rat liver; Cellular Location: Cell membrane, Multi-pass membrane protein.
This gene is located within the Smith-Magenis syndrome region on chromosome 17. It encodes a protein of unknown function.
Specifications
SLC47A1 | |
Polyclonal | |
Unconjugated | |
Slc47a1 | |
1300013J15Rik; AI036982; FLJ10847; H+/organic cation antiporter variant 1; H+/organic cation antiporter variant 2; hMATE-1; MATE1; MATE-1; MGC64822; mMATE1; mMATE-1; multidrug and toxin extrusion 1; multidrug and toxin extrusion protein 1; RGD1311123; rMATE-1; SLC47A1; solute carrier family 47 (multidrug and toxin extrusion), member 1; solute carrier family 47 member 1; solute carrier family 47, member 1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
360539, 55244 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.98 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q5I0E9, Q96FL8 | |
Slc47a1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 460-550 of human SLC47A1 (NP_0607122). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction