Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC47A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537990
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537990 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537990 Supplier Invitrogen™ Supplier No. PA537990
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

S47A2 belongs to a family of transporters involved in excretion of toxic electrolytes, both endogenous and exogenous, through urine and bile. This transporter family shares homology with the bacterial MATE (multidrug and toxin extrusion) protein family responsible for drug resistance. This gene is one of two members of the MATE transporter family located near each other on chromosome 17.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC47A2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Slc47a2
Gene Accession No. Q86VL8
Gene Alias 4933429E10Rik; FLJ31196; H(+)/organic cation antiporter kidney-specific; hMATE-2; kidney-specific H(+)/organic cation antiporter; Mate2; MATE-2; MATE2-B; MATE2K; MATE2-K; mMATE-2; multidrug and toxin extrusion protein 2; SLC47A2; solute carrier family 47 (multidrug and toxin extrusion), member 2; solute carrier family 47 member 2; solute carrier family 47, member 2
Gene Symbols Slc47a2
Host Species Goat
Immunogen Peptide with sequence C-QRAESTATRPGPEK, from the internal region near the C-terminus of SLC47A2 (aa 511-525).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146802
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.